Learn More
Thermo Scientific Chemicals 3'-Deoxythymidine
Product Code. 15407069
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
Product Code. 15407069
Brand: Thermo Scientific Chemicals J65765.03
Request Bulk or Custom Format
CAS: 3416-05-5 | C10H14N2O4 | 226.23 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3416-05-5
Molecular Formula C10H14N2O4
Molecular Weight (g/mol) 226.23
MDL Number MFCD00010570
InChI Key XKKCQTLDIPIRQD-JGVFFNPUSA-N
Synonym 3'-deoxythymidine, 2',3'-dideoxythymidine, 1-2r,5s-5-hydroxymethyl tetrahydrofuran-2-yl-5-methylpyrimidine-2,4 1h,3h-dione, dideoxythymidine, thymidine, 3'-deoxy, desoxythymidin, 1-2r,5s-5-hydroxymethyl tetrahydrofuran-2-yl-5-methyl-pyrimidine-2,4-dione, ddt & gm-csf
PubChem CID 65119
IUPAC Name 1-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione
SMILES CC1=CN([C@H]2CC[C@@H](CO)O2)C(=O)NC1=O
Specifications
Melting Point 155°C to 156°C
Color White to Yellow
Quantity 1 g
Beilstein 21884
Formula Weight 226.23
Physical Form Powder
Chemical Name or Material 3'-Deoxythymidine