missing translation for 'onlineSavingsMsg'
Learn More

3-Deoxyglucosone, TRC

Product Code. 30007208
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007208 5 mg 5mg
30010178 25 mg 25mg
30012710 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007208 Supplier Toronto Research Chemicals Supplier No. TRCD2391505MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4084-27-9
Molecular Formula C6 H10 O5
Molecular Weight (g/mol) 162.14
Synonym D-erythro-Hexos-2-ulose, 3-deoxy-, D-erythro-Hexosulose, 3-deoxy- (6CI,7CI,8CI), 3-Deoxy-D-erythro-hexos-2-ulose, 2-keto-3-Deoxyglucose, 3-Deoxy-D-erythro-hexosulose, 3-Deoxy-D-glucosone, 3-Deoxyglucosone, (4S,5R)-4,5,6-Trihydroxy-2-oxohexanal
IUPAC Name (4S,5R)-4,5,6-trihydroxy-2-oxohexanal
SMILES OC[C@@H](O)[C@@H](O)CC(=O)C=O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C6H10O5/c7-2-4(9)1-5(10)6(11)3-8/h2,5-6,8,10-11H,1,3H2/t5-,6+/m0/s1
Formula Weight 162.0528
Percent Purity >95%
Chemical Name or Material 3-Deoxyglucosone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.