Learn More
3-(Cyclohexylamino)-1-propanesulfonic-d17 Acid, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.1g
Specifications
Specifications
| Chemical Name or Material | 3-(Cyclohexylamino)-1-propanesulfonic Acid-D17 |
| CAS | 1219804-15-5 |
| Molecular Formula | C9 D17 H2 N O3 S |
| Quantity | 0.1 g |
| Synonym | 3-(Cyclohexylamino)-1-propanesulfonic-d17 Acid, 3-(Cyclohexylamino)-1-propanesulfonic Acid-D17, CAPS-d17, N-(Cyclohexyl-d11)-3-aminopropane-1,1,2,2,3,3-d6-sulfonic acid |
| InChI Formula | InChI=1 S/C9H19NO3S/c11-14(12,13)8-4-7-10-9-5-2-1-3-6-9/h9-10 H,1-8H2,(H,11,12,13)/i1D2,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9 D |
| SMILES | [2 H]C([2 H])(NC1([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C1([2 H])[2 H])C([2 H])([2 H])C([2 H])([2 H])S(=O)(=O)O |
| IUPAC Name | 1,1,2,2,3,3-hexadeuterio-3-[(1,2,2,3,3,4,4,5,5,6,6-undecadeuteriocyclohexyl)amino]propane-1-sulfonic acid |
| Molecular Weight (g/mol) | 238.4218 |
| Formula Weight | 238.2153 g/mol |
| Show More |