Learn More
3-Cyclohexyl-1-propyne, 97%
Description
3-Cyclohexyl-1-propyne, is used as a pharmaceutical intermediate. It can be used to produce (5-bromo-4,4,5,5-tetrafluoro-pent-2-enyl)-cyclohexane at the temperature of 40°C. It will need reagents (NH4)2S2O8, HCO2Na·2H2O and solvent dimethylformamide with the reaction time of 3 hours. It is also used as organic synthesis intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3-Cyclohexyl-1-propyne, is used as a pharmaceutical intermediate. It can be used to produce (5-bromo-4,4,5,5-tetrafluoro-pent-2-enyl)-cyclohexane at the temperature of 40°C. It will need reagents (NH4)2S2O8, HCO2Na·2H2O and solvent dimethylformamide with the reaction time of 3 hours. It is also used as organic synthesis intermediates.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications
Specifications
| Density | 0.845 |
| Boiling Point | 157°C to 158°C |
| Flash Point | 35°C (95°F) |
| Refractive Index | 1.462 |
| Quantity | 5 g |
| UN Number | UN3295 |
| Beilstein | 1849402 |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 122.21 |
| Percent Purity | 97% |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.