missing translation for 'onlineSavingsMsg'
Learn More

(3-Chloropropyl)triethoxysilane, 97+%

Product Code. 10579023 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
25g
5g
This item is not returnable. View return policy

Product Code. 10579023

Brand: Thermo Scientific Acros 391890010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5089-70-3 | C9H21ClO3Si | 240.8 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5089-70-3
Molecular Formula C9H21ClO3Si
Molecular Weight (g/mol) 240.8
MDL Number MFCD00018985
InChI Key KSCAZPYHLGGNPZ-UHFFFAOYSA-N
Synonym 3-chloropropyl triethoxysilane, 3-chloropropyl triethoxy silane, silane, 3-chloropropyl triethoxy, unii-x7rb20518m, triethoxy gamma-chloropropyl silane, gamma-chloropropyltriethoxysilane, triethoxy .gamma.-chloropropyl silane, chloropropyl triethoxysilane, dynasylan cpteo
PubChem CID 78771
IUPAC Name 3-chloropropyl(triethoxy)silane
SMILES CCO[Si](CCCCl)(OCC)OCC

Specifications

CAS Min % 97.0
CAS Max % 100.0
Color Yellow
Density 1.0040g/mL
Boiling Point 230.0°C
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Linear Formula Cl(CH2)3Si(OCH2CH3)
Refractive Index 1.4190 to 1.4210 (20°C, 589nm)
Quantity 1 kg
Specific Gravity 1.004
Solubility Information Solubility in water: insoluble
Formula Weight 240.8
Percent Purity 97+%
Physical Form Liquid
Chemical Name or Material (3-Chloropropyl)triethoxysilane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.