Learn More
3-Chlorodiphenylamine, TRC
Specifications
Specifications
| CAS | 101-17-7 |
| Molecular Formula | C12 H10 Cl N |
| Quantity | 1 g |
| Synonym | 3-Chloro-N-phenyl-benzenamine, N-(3-Chlorophenyl)-N-phenylamine, N-(3-Chlorophenyl)aniline, N-(m-Chlorophenyl)aniline, Phenyl(m-chlorophenyl)amine, m-Chlorodiphenylamine |
| InChI Formula | InChI=1S/C12H10ClN/c13-10-5-4-8-12(9-10)14-11-6-2-1-3-7-11/h1-9,14H |
| SMILES | Clc1cccc(Nc2ccccc2)c1 |
| IUPAC Name | 3-chloro-N-phenylaniline |
| Molecular Weight (g/mol) | 203.67 |
| Formula Weight | 203.0502 |
| Chemical Name or Material | 3-Chlorodiphenylamine |