missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-6-methoxybenzo[b]thiophene-2-carbonyl chloride, Thermo Scientific™

Product Code. 11315439
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
11315439 250 mg 250mg
11325439 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11315439

Brand: Thermo Scientific Alfa Aesar H50419.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75998-29-7
Molecular Formula C10H6Cl2O2S
Molecular Weight (g/mol) 261.116
MDL Number MFCD00448907
InChI Key DTMXTCKEPQWRPO-UHFFFAOYSA-N
Synonym 3-chloro-6-methoxybenzo b thiophene-2-carbonyl chloride, benzo b thiophene-2-carbonyl chloride, 3-chloro-6-methoxy, acmc-20an6b, aronis24536, 3-chloro-6-methoxy-benzo b thiophene-2-carbonyl chloride, 3-chloro-6-methoxybenzo-b thiophene-2-carbonyl chloride, 6-methoxy-3-chlorobenzo b thiophene-2-carbonyl chloride, 6-methoxy-3-chloro-benzo b-thiophene-2-carbonyl chloride, benzo b thiophene-2-carbonylchloride, 3-chloro-6-methoxy
PubChem CID 12594638
IUPAC Name 3-chloro-6-methoxy-1-benzothiophene-2-carbonyl chloride
SMILES COC1=CC2=C(C=C1)C(=C(S2)C(=O)Cl)Cl

Specifications

Quantity 250 mg
UN Number UN3261
Formula Weight 261.13
Chemical Name or Material 3-Chloro-6-methoxybenzo[b]thiophene-2-carbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.