Learn More
3-Chloro-5-(pentafluorothio)benzaldehyde, 97%, Thermo Scientific™
Product Code. 15446067
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15446067 250 mg 250mg
15436067 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15446067 Supplier Thermo Scientific Alfa Aesar Supplier No. H33248.MD
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1240257-88-8
Molecular Formula C7H4ClF5OS
Molecular Weight (g/mol) 266.61
MDL Number MFCD16652412
InChI Key AHEFJQKJBDJLSO-UHFFFAOYSA-N
Synonym 3-chloro-5-pentafluorosulfur benzaldehyde, 3-chloro-5-pentafluoro-??-sulfanyl benzaldehyde, 3-chloro-5-pentafluorothio benzaldehyde, 3-chloro-5-5-pentafluorothio benzaldehyde
PubChem CID 66523453
IUPAC Name 3-chloro-5-(pentafluoro-$l^{6}-sulfanyl)benzaldehyde
SMILES FS(F)(F)(F)(F)C1=CC(Cl)=CC(C=O)=C1
Specifications
Quantity 250 mg
Sensitivity Air sensitive
Formula Weight 266.61
Percent Purity 97%
Chemical Name or Material 3-Chloro-5-(pentafluorothio)benzaldehyde