Learn More
3-Chloro-4-nitroaniline, 95%
Product Code. 11314666
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11314666 1 g 1g
11304666 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11314666 Supplier Thermo Scientific Alfa Aesar Supplier No. H32581.03
 Restricted Item
In Stock
Add to basket
CAS: 825-41-2 | C6H5ClN2O2 | 172.568 g/mol
3-Chloro-4-nitroaniline is used as a pharmaceutical and medical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Chloro-4-nitroaniline is used as a pharmaceutical and medical intermediate.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 825-41-2
Molecular Formula C6H5ClN2O2
Molecular Weight (g/mol) 172.568
MDL Number MFCD00085922
InChI Key LDSIOPGMLLPSSR-UHFFFAOYSA-N
Synonym benzenamine, 3-chloro-4-nitro, aniline, 3-chloro-4-nitro, 3-chlor-4-nitroanilin, 3-chloro-4-nitrobenzenamine, zlchem 908, pubchem9594, acmc-209ppd, 2-chloro-4-aminonitrobenzene, 4-amino-2-chloronitrobenzene, 3-chloro-4-nitro-phenylamine
PubChem CID 69996
IUPAC Name 3-chloro-4-nitroaniline
SMILES C1=CC(=C(C=C1N)Cl)[N+](=O)[O-]
Spécification
Melting Point 157°C to 161°C
Quantity 1 g
UN Number UN2811
Solubility Information Insoluble in water.
Formula Weight 172.57
Percent Purity 95%
Chemical Name or Material 3-Chloro-4-nitroaniline
RUO – Research Use Only
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.