Learn More
3’-Chloro-4’-hydroxyacetophenone, TRC
Specifications
Specifications
| Chemical Name or Material | 3'-Chloro-4'-hydroxyacetophenone |
| CAS | 2892-29-7 |
| Molecular Formula | C8 H7 Cl O2 |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C8H7ClO2/c1-5(10)6-2-3-8(11)7(9)4-6/h2-4,11H,1H3 |
| SMILES | CC(=O)c1ccc(O)c(Cl)c1 |
| IUPAC Name | 1-(3-chloro-4-hydroxyphenyl)ethanone |
| Molecular Weight (g/mol) | 170.59 |
| Formula Weight | 170.0135 |