Learn More
3-Chloro-4-(3-fluorobenzyloxy)aniline, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Chloro-4-(3-fluorobenzyloxy)aniline |
| CAS | 202197-26-0 |
| Molecular Formula | C13 H11 Cl F N O |
| Quantity | 1 g |
| Synonym | 3-Chloro-4-[(3-fluorophenyl)methoxy]benzenamine, 3-Chloro-4-[(3-fluorobenzyl)oxy]aniline, 3-Chloro-4-[(3-fluorophenyl)methoxy]aniline, 4-(3-Fluorobenzyloxy)-3-chloroaniline, [3-Chloro-4-(3-fluorobenzyloxy)phenyl]amine, GW 397339X, Lapatinib related, GW572016X, GW562778B, GW684529A, GW680445X, GW621412X, GW671286X, GW607864X, GW560231C, GW574783C, GW680835A, GW696252X, Lapatinib related, GW 397339X, GW 359812X, GR 211784X, GW 404735X, GW 443025X, GSK 203025A, GW 394943X, GW 834380, GSK 726595A |
| InChI Formula | InChI=1S/C13H11ClFNO/c14-12-7-11(16)4-5-13(12)17-8-9-2-1-3-10(15)6-9/h1-7H,8,16H2 |
| SMILES | Nc1ccc(OCc2cccc(F)c2)c(Cl)c1 |
| IUPAC Name | 3-chloro-4-[(3-fluorophenyl)methoxy]aniline |
| Molecular Weight (g/mol) | 251.68 |
| Formula Weight | 251.0513 |