Learn More
3-Chloro-2-fluorobenzeneboronic acid, 97%
Product Code. 15439417
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 15439417
Brand: Thermo Scientific Alfa Aesar H52665.06
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
CAS: 352535-82-1 | C6H5BClFO2 | 174.36 g/mol
3-Chloro-2-fluorobenzeneboronic acid, Is used inorganic synthesis, catalysis, analytical chemistry and biological systems.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Chloro-2-fluorobenzeneboronic acid, Is used inorganic synthesis, catalysis, analytical chemistry and biological systems.
Solubility
Insoluble in water. Soluble in organic solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 352535-82-1
Molecular Formula C6H5BClFO2
Molecular Weight (g/mol) 174.36
MDL Number MFCD05664224
InChI Key SUYRGLRWMPEARP-UHFFFAOYSA-N
Synonym 3-chloro-2-fluorophenyl boronic acid, 3-chloro-2-fluorobenzeneboronic acid, boronic acid, 3-chloro-2-fluorophenyl, boronic acid,b-3-chloro-2-fluorophenyl, acmc-209ifa, 3-chloro-2-fluorophenyl boranediol, 3-chloro-2-fluorophenyl boronicacid, 2-fluoro-3-chlorophenylboronic acid, 3-chloro-2-fluorophenylboronic acid
PubChem CID 16217158
IUPAC Name (3-chloro-2-fluorophenyl)boronic acid
SMILES OB(O)C1=CC=CC(Cl)=C1F
Specifications
Melting Point 240°C to 242°C
Quantity 5 g
Solubility Information Insoluble in water. Soluble in organic solvents.
Formula Weight 174.37
Percent Purity 97%
Chemical Name or Material 3-Chloro-2-fluorobenzeneboronic acid