Learn More
3-Chloro-2,5-difluoropyridine, 97%
Product Code. 15445387
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15445387
Brand: Thermo Scientific Alfa Aesar H32640.MD
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
CAS: 851179-00-5 | C5H2ClF2N | 149.525 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 851179-00-5
Molecular Formula C5H2ClF2N
Molecular Weight (g/mol) 149.525
MDL Number MFCD09998887
InChI Key ATKDHGGWYZLPPJ-UHFFFAOYSA-N
Synonym 3-chloro-2,5-difluoro-pyridine, pyridine, 3-chloro-2,5-difluoro, ksc654c8n
PubChem CID 19371877
IUPAC Name 3-chloro-2,5-difluoropyridine
SMILES C1=C(C=NC(=C1Cl)F)F
Specifications
Refractive Index 1.48
Quantity 250 mg
UN Number UN2924
Formula Weight 149.53
Percent Purity 97%
Chemical Name or Material 3-Chloro-2,5-difluoropyridine