Learn More
3-Chloro-1-indan-5-yl-propan-1-one, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Chloro-1-indan-5-yl-propan-1-one |
| CAS | 39105-39-0 |
| Molecular Formula | C12 H13 Cl O |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C12H13ClO/c13-7-6-12(14)11-5-4-9-2-1-3-10(9)8-11/h4-5,8H,1-3,6-7H2 |
| SMILES | ClCCC(=O)c1ccc2CCCc2c1 |
| IUPAC Name | 3-chloro-1-(2,3-dihydro-1H-inden-5-yl)propan-1-one |
| Molecular Weight (g/mol) | 208.68 |
| Formula Weight | 208.0655 |