missing translation for 'onlineSavingsMsg'
Learn More

Chlorodibromoacetaldehyde (>90%), TRC

Product Code. 30364027
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30364027 250 mg -
30367442 1 g -
30354820 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30364027 Supplier Toronto Research Chemicals Supplier No. TRCC370985250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64316-11-6
Molecular Formula C2 H Br2 Cl O
Molecular Weight (g/mol) 236.29
Synonym 2,2-Dibromo-2-chloro-acetaldehyde, Chlorodibromoacetaldehyde, Dibromochloroacetaldehyde, Dibromochloro-acetaldehyde
IUPAC Name 2,2-dibromo-2-chloroacetaldehyde
SMILES ClC(Br)(Br)C=O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C2HBr2ClO/c3-2(4,5)1-6/h1H
Formula Weight 233.808
Chemical Name or Material Chlorodibromoacetaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.