missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(3-Bromopyridin-2-yl)methan-d2-ol, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | (3-Bromopyridin-2-yl)methanol-D2 |
| CAS | 2170188-53-9 |
| Molecular Formula | C6 D2 H4 Br N O |
| Quantity | 0.1 g |
| Synonym | 3-Bromo-2-pyridinemethanol-α,α-d2, (3-Bromopyridin-2-yl)methanol-D2, 3-Bromo-2-(hydroxymethyl-d2)pyridine, (3-Bromopyridin-2-yl)methan-d2-ol |
| InChI Formula | InChI=1 S/C6H6BrNO/c7-5-2-1-3-8-6(5)4-9/h1-3,9 H,4H2/i4D2 |
| SMILES | [2 H]C([2 H])(O)c1ncccc1Br |
| IUPAC Name | (3-bromopyridin-2-yl)-dideuteriomethanol |
| Molecular Weight (g/mol) | 190.0343 |
| Formula Weight | 188.9758 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction