Learn More
3-Bromophenyl isothiocyanate, 97%, Thermo Scientific™
Product Code. 10507874
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10507874 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10507874 Supplier Thermo Scientific Maybridge Supplier No. S04141DA
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
CAS 2131-59-1
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C7H4BrNS
Linear Formula BrC6H4NCS
MDL Number MFCD00004803
Quantity 1 g
Synonym 3-bromophenyl isothiocyanate, m-bromophenyl isothiocyanate, benzene, 1-bromo-3-isothiocyanato, isothiocyanic acid, m-bromophenyl ester, 3-bromophenylisothiocyanate, 1-bromo-3-isothiocyanato-benzene, acmc-1chhg, m-bromphenylisothiocyanat, zmgmghnoacsmqn-uhfffaoysa, #
InChI Key ZMGMGHNOACSMQN-UHFFFAOYSA-N
SMILES C1=CC(=CC(=C1)Br)N=C=S
IUPAC Name 1-bromo-3-isothiocyanatobenzene
Molecular Weight (g/mol) 214.08
PubChem CID 16481
Formula Weight 214.09
Percent Purity 97%
Chemical Name or Material 3-Bromophenyl isothiocyanate