missing translation for 'onlineSavingsMsg'
Learn More

3-(Bromomethyl)-Alpha,Alpha,5-trimethyl-benzeneacetonitrile, TRC

Product Code. 30336310
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30336310 100 mg 100mg
30332998 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30336310 Supplier Toronto Research Chemicals Supplier No. TRCB685860100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(Bromomethyl)-alpha,alpha,5-trimethyl-benzeneacetonitrile
CAS 120512-36-9
Molecular Formula C12H14BrN
Quantity 100 mg
Synonym 2-(3-Bromomethyl-5-methylphenyl)-2-methylpropionitrile
InChI Formula InChI=1S/C12H14BrN/c1-9-4-10(7-13)6-11(5-9)12(2,3)8-14/h4-6H,7H2,1-3H3
SMILES Cc1cc(CBr)cc(c1)C(C)(C)C#N
IUPAC Name 2-[3-(bromomethyl)-5-methylphenyl]-2-methylpropanenitrile
Molecular Weight (g/mol) 252.15
Formula Weight 251.031
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.