Learn More
3-Bromo Labetalol Hydrochloride Salt, TRC
Product Code. 30336400
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30336400 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30336400 Supplier Toronto Research Chemicals Supplier No. TRCB6887865MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Bromo Labetalol Hydrochloride Salt
CAS 2445226-37-7
Molecular Formula C19H23BrN2O3 • HCl
Quantity 5 mg
Synonym Benzamide, 3-bromo-2-hydroxy-5-[1-hydroxy-2-[(1-methyl-3-phenylpropyl)amino]ethyl]-, hydrochloride (1:1) (ACI)
InChI Formula InChI=1S/C19H23BrN2O3.ClH/c1-12(7-8-13-5-3-2-4-6-13)22-11-17(23)14-9-15(19(21)25)18(24)16(20)10-14;/h2-6,9-10,12,17,22-24H,7-8,11H2,1H3,(H2,21,25);1H
SMILES O=C(N)C1=CC(C(O)CNC(C)CCC2=CC=CC=C2)=CC(Br)=C1O.Cl
IUPAC Name 3-bromo-2-hydroxy-5-(1-hydroxy-2-((4-phenylbutan-2-yl)amino)ethyl)benzamide hydrogen chloride
Molecular Weight (g/mol) 443.76
Formula Weight 442.07