Learn More
3-Bromo-6-pyridinecarbonitrile, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Bromo-6-pyridinecarbonitrile |
| CAS | 97483-77-7 |
| Molecular Formula | C6 H3 Br N2 |
| Quantity | 1 g |
| Synonym | 2-Pyridinecarbonitrile, 5-bromo-, 5-Bromo-2-pyridinecarbonitrile, 2-Cyano-5-bromopyridine, 3-Bromo-6-cyanopyridine, 3-Bromo-6-pyridinecarbonitrile, 5-Bromo-2-cyanopyridine, 5-Bromopicolinonitrile, 5-Bromopyridine-2-carbonitrile |
| InChI Formula | InChI=1S/C6H3BrN2/c7-5-1-2-6(3-8)9-4-5/h1-2,4H |
| SMILES | Brc1ccc(nc1)C#N |
| IUPAC Name | 5-bromopyridine-2-carbonitrile |
| Molecular Weight (g/mol) | 183.01 |
| Formula Weight | 181.948 |