missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-(trifluoromethoxy)benzaldehyde, 97%, Thermo Scientific™

Product Code. 11495960
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495960 1 g 1g
11405970 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11495960 Supplier Thermo Scientific Alfa Aesar Supplier No. H26544.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886498-07-3
Molecular Formula C8H4BrF3O2
Molecular Weight (g/mol) 269.017
MDL Number MFCD04116040
InChI Key ALIVUZVZCLVJQN-UHFFFAOYSA-N
Synonym 3-bromo-5-trifluoromethoxy benzaldehyde, acmc-209qvy, 3-bromo-5-trifluoromethoxy-benzaldehyde
PubChem CID 5006711
IUPAC Name 3-bromo-5-(trifluoromethoxy)benzaldehyde
SMILES C1=C(C=C(C=C1OC(F)(F)F)Br)C=O

Specifications

Refractive Index 1.499
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 269.01
Percent Purity 97%
Chemical Name or Material 3-Bromo-5-(trifluoromethoxy)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.