Learn More
3-Bromo-5-methylanisole, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Bromo-5-methylanisole |
| CAS | 29578-83-4 |
| Molecular Formula | C8 H9 Br O |
| Quantity | 2 g |
| InChI Formula | InChI=1S/C8H9BrO/c1-6-3-7(9)5-8(4-6)10-2/h3-5H,1-2H3 |
| SMILES | COc1cc(C)cc(Br)c1 |
| IUPAC Name | 1-bromo-3-methoxy-5-methylbenzene |
| Molecular Weight (g/mol) | 201.06 |
| Formula Weight | 199.9837 |