Learn More
3-Bromo-4-nitrophenol, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Bromo-4-nitrophenol |
| CAS | 5470-65-5 |
| Molecular Formula | C6 H4 Br N O3 |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C6H4BrNO3/c7-5-3-4(9)1-2-6(5)8(10)11/h1-3,9H |
| SMILES | Oc1ccc(c(Br)c1)[N+](=O)[O-] |
| IUPAC Name | 3-bromo-4-nitrophenol |
| Molecular Weight (g/mol) | 218 |
| Formula Weight | 216.9375 |