Learn More
3-Bromo-4-methoxybenzaldehyde, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Bromo-4-methoxybenzaldehyde |
| CAS | 34841-06-0 |
| Molecular Formula | C8H7BrO2 |
| Quantity | 25 g |
| Synonym | 3-Bromo-4-methoxybenzaldehyde, 3-Bromo-p-anisaldehyde, NSC 158162 |
| InChI Formula | InChI=1S/C8H7BrO2/c1-11-8-3-2-6(5-10)4-7(8)9/h2-5H,1H3 |
| SMILES | COc1ccc(C=O)cc1Br |
| IUPAC Name | 3-bromo-4-methoxybenzaldehyde |
| Molecular Weight (g/mol) | 215.04 |
| Formula Weight | 213.9629 |