Learn More
(3-Bromo-4-hydroxyphenyl)(2-ethyl-3-benzofuranyl)-methanone, TRC
Specifications
Specifications
| Chemical Name or Material | (3-Bromo-4-hydroxyphenyl)(2-ethyl-3-benzofuranyl)-methanone |
| CAS | 94729-09-6 |
| Molecular Formula | C17H13BrO3 |
| Quantity | 250 mg |
| Synonym | 3-Bromo-4-hydroxyphenyl 2-ethyl-3-benzofuranyl Ketone,, (3-Bromo-4-hydroxyphenyl)(2-ethylbenzofuran-3-yl)methanone, Bromobenzarone, Benzbromarone Impurity A |
| InChI Formula | InChI=1S/C17H13BrO3/c1-2-14-16(11-5-3-4-6-15(11)21-14)17(20)10-7-8-13(19)12(18)9-10/h3-9,19H,2H2,1H3 |
| SMILES | CCc1oc2ccccc2c1C(=O)c3ccc(O)c(Br)c3 |
| IUPAC Name | (3-bromo-4-hydroxyphenyl)-(2-ethyl-1-benzofuran-3-yl)methanone |
| Molecular Weight (g/mol) | 345.19 |
| Formula Weight | 344.0048 |