missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-hydroxy-5-methoxycinnamic acid, 97%

Product Code. 11400471
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11400471 1 g 1g
11410471 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11400471 Supplier Thermo Scientific Alfa Aesar Supplier No. B20014.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 6948-33-0 | C10H8BrO4 | 272.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6948-33-0
Molecular Formula C10H8BrO4
Molecular Weight (g/mol) 272.08
MDL Number MFCD00017334
InChI Key NGBVBJQUJJKRGF-NSCUHMNNSA-M
Synonym 3-3-bromo-4-hydroxy-5-methoxyphenyl acrylic acid, 5-bromoferulic acid, 2e-3-3-bromo-4-hydroxy-5-methoxyphenyl prop-2-enoic acid, 3-3-bromo-4-hydroxy-5-methoxyphenyl prop-2-enoic acid, 3-bromo-4-hydroxy-5-methoxycinnamic acid, 2e-3-3-bromo-4-hydroxy-5-methoxyphenyl acrylic acid, 2-propenoic acid,3-3-bromo-4-hydroxy-5-methoxyphenyl, e-3-3-bromo-4-hydroxy-5-methoxy-phenyl prop-2-enoic acid, e-3-3-bromo-4-hydroxy-5-methoxyphenyl prop-2-enoic acid
PubChem CID 735789
SMILES COC1=CC(\C=C\C([O-])=O)=CC(Br)=C1O

Specifications

Melting Point 255°C to 257°C
Odor Odorless
Quantity 1 g
IUPAC Name (2E)-3-(3-bromo-4-hydroxy-5-methoxyphenyl)prop-2-enoate
Formula Weight 273.09
Percent Purity 97%
Chemical Name or Material 3-Bromo-4-hydroxy-5-methoxycinnamic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.