Learn More
3-Bromo-4-(dibromomethyl)-5-hydroxy-2(5H)-furanone, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Bromo-4-(dibromomethyl)-5-hydroxy-2(5H)-furanone |
| CAS | 132059-53-1 |
| Molecular Formula | C5 H3 Br3 O3 |
| Quantity | 1 mg |
| Synonym | BMX 3 |
| InChI Formula | InChI=1S/C5H3Br3O3/c6-2-1(3(7)8)4(9)11-5(2)10/h3-4,9H |
| SMILES | OC1OC(=O)C(=C1C(Br)Br)Br |
| IUPAC Name | 4-bromo-3-(dibromomethyl)-2-hydroxy-2H-furan-5-one |
| Molecular Weight (g/mol) | 350.79 |
| Formula Weight | 347.7632 |