Learn More
3-Bromo-3-methyl-2-butanone, TRC
Product Code. 30330618
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30330618 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30330618 Supplier Toronto Research Chemicals Supplier No. TRCB67847350MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Bromo-3-methyl-2-butanone
CAS 2648-71-7
Molecular Formula C5H9OBr
Quantity 50 mg
InChI Formula InChI=1S/C5H9BrO/c1-4(7)5(2,3)6/h1-3H3
SMILES CC(=O)C(C)(C)Br
IUPAC Name 3-bromo-3-methylbutan-2-one
Molecular Weight (g/mol) 165.02
Formula Weight 163.9837