missing translation for 'onlineSavingsMsg'
Learn More

3-Boc-3-(methylamino)azetidine hydrochloride, 95%

Product Code. 15442498
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15442498

Brand: Thermo Scientific Alfa Aesar H56060.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 577777-20-9 | C9H18N2O2 | 186.26 g/mol

It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Solubility
Soluble in water. (5 g/100ml)

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 577777-20-9
Molecular Formula C9H18N2O2
Molecular Weight (g/mol) 186.26
MDL Number MFCD12963843,MFCD08061963
InChI Key LHUFQYUQIUJJIB-UHFFFAOYSA-N
Synonym tert-butyl azetidin-3-yl methyl carbamate, 3-boc-3-methylaminoazetidine, 3-n-boc-n-methylamino-azetidine, tert-butyl n-azetidin-3-yl-n-methylcarbamate, carbamic acid, 3-azetidinylmethyl-, 1,1-dimethylethyl ester, 3-boc-3-methylamino azetidine hydrochloride, 3-boc-methylamino azetidine, 3-n-boc-methylamino azatidine, 3-boc-3-methylaminoazetidine hcl, tert-butyl azetidin-3-yl-methylcarbamate
PubChem CID 23033645
IUPAC Name tert-butyl N-(azetidin-3-yl)-N-methylcarbamate
SMILES CN(C1CNC1)C(=O)OC(C)(C)C

Specifications

Solubility Information Soluble in water. (5g/100ml)
Quantity 1 g
Formula Weight 222.72
Percent Purity 95%
Chemical Name or Material 3-Boc-3-(methylamino)azetidine hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.