Learn More
3-Benzyloxyphenylacetonitrile, 97%, Thermo Scientific™
Product Code. 11324116
Change view
Click to view available options
Quantity:
500 mg
2 g
Unit Size:
2g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11324116 2 g 2g
11314116 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11324116 Supplier Thermo Scientific Alfa Aesar Supplier No. L19241.04
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 20967-96-8
Molecular Formula C15H13NO
Molecular Weight (g/mol) 223.28
MDL Number MFCD00016396
InChI Key CKZFVIPFANUBDW-UHFFFAOYSA-N
Synonym 3-benzyloxyphenylacetonitrile, 2-3-benzyloxy phenyl acetonitrile, 3-benzyloxybenzyl cyanide, 2-3-phenylmethoxyphenyl acetonitrile, 2-3-phenylmethoxy phenyl ethanenitrile, maybridge3_000453, 3-benzyloxyphenyl acetonitrile, 3-benzyloxy-phenyl acetonitrile, 3-benzyloxyphenyl-acetonitrile
PubChem CID 519831
IUPAC Name 2-(3-phenylmethoxyphenyl)acetonitrile
SMILES N#CCC1=CC(OCC2=CC=CC=C2)=CC=C1
Specifications
Melting Point 47°C to 50°C
Boiling Point 188°C to 190°C (3 mmHg)
Quantity 2 g
Beilstein 2585802
Formula Weight 223.28
Percent Purity 97%
Chemical Name or Material 3-Benzyloxyphenylacetonitrile