missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Aminobenzoic-2,4,5,6-d4 Acid, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.10g
Specifications
Specifications
| Chemical Name or Material | 3-Aminobenzoic-d4 Acid |
| CAS | 78399-79-8 |
| Molecular Formula | C7 D4 H3 N O2 |
| Quantity | 0.1 g |
| Synonym | Benzoic-2,3,4,6-d4 acid, 5-amino- (9 CI), 3-Aminobenzoic-2,4,5,6-d4 Acid, 3-Aminobenzoic Acid-2,4,5,6-D4, 3-Aminobenzoic Acid-d4 |
| InChI Formula | InChI=1 S/C7H7NO2/c8-6-3-1-2-5(4-6)7(9)10/h1-4 H,8H2,(H,9,10)/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c(N)c([2 H])c(C(=O)O)c1[2 H] |
| IUPAC Name | 3-amino-2,4,5,6-tetradeuteriobenzoic acid |
| Molecular Weight (g/mol) | 141.1606 |
| Formula Weight | 141.0728 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction