Learn More
3-amino-6-cyanopyridazine, TRC
Specifications
Specifications
| Chemical Name or Material | 3-amino-6-cyanopyridazine |
| CAS | 340759-46-8 |
| Molecular Formula | C5 H4 N4 |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C5H4N4/c6-3-4-1-2-5(7)9-8-4/h1-2H,(H2,7,9) |
| SMILES | N=C1NN=C(C=C1)C#N |
| IUPAC Name | 6-imino-1H-pyridazine-3-carbonitrile |
| Molecular Weight (g/mol) | 120.112 |
| Formula Weight | 120.0436 |