Learn More
3-Amino-5-bromo-2-methoxypyridine, 96%, Thermo Scientific Chemicals
Product Code. 15410628
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15410628 250 mg 250mg
15490618 1 g 1g
15400628 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15410628 Supplier Thermo Scientific Alfa Aesar Supplier No. H54299.MD
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 884495-39-0 | Molecular Formula: C6H7BrN2O | Molecular Weight: 203.039 g/mol
3-Amino-5-bromo-2-methoxypyridine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Amino-5-bromo-2-methoxypyridine is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 884495-39-0
Molecular Formula C6H7BrN2O
Molecular Weight (g/mol) 203.039
MDL Number MFCD07368887
InChI Key HJOOFLFWIISCAI-UHFFFAOYSA-N
Synonym 3-amino-5-bromo-2-methoxypyridine, 5-bromo-2-methoxy-3-pyridinamine, 3-amino-5-bromopyridin-2-yl methyl ether, 5-bromo-2-methoxy-3-aminopyridine, 5-bromo-2-methoxy-3-cyanopyridine, 5-bromo-2-methyloxy-3-pyridinamine, 3-pyridinamine,5-bromo-2-methoxy, 5-bromo-2-methoxy-pyridin-3-ylamine, pubchem17164, acmc-209qtg
PubChem CID 44754869
IUPAC Name 5-bromo-2-methoxypyridin-3-amine
SMILES COC1=NC=C(C=C1N)Br
Specifications
Melting Point 53°C to 55°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 203.04
Percent Purity 96%
Chemical Name or Material 3-Amino-5-bromo-2-methoxypyridine