Learn More
3-Amino-4-chlorobenzoic Acid, TRC
Product Code. 30311559
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30311559 5 g 5g
30325905 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30311559 Supplier Toronto Research Chemicals Supplier No. TRCA6177455G
 Restricted Item
Estimated Shipment: 04-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Spezifikation
Chemical Name or Material 3-Amino-4-chlorobenzoic Acid
CAS 2840-28-0
Molecular Formula C7H6ClNO2
Quantity 5 g
InChI Formula InChI=1S/C7H6ClNO2/c8-5-2-1-4(7(10)11)3-6(5)9/h1-3H,9H2,(H,10,11)
SMILES Nc1cc(ccc1Cl)C(=O)O
IUPAC Name 3-amino-4-chlorobenzoic acid
Molecular Weight (g/mol) 171.58
Formula Weight 171.0087