Learn More
3-Amino-2-naphthol, 97%
Product Code. 11460418
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460418 25 g 25g
11440418 1 g 1g
11450418 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11460418 Supplier Thermo Scientific Alfa Aesar Supplier No. B24570.14
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
CAS: 5417-63-0 | C10H9NO | 159.188 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5417-63-0
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.188
MDL Number MFCD00004113
InChI Key ZHVPTERSBUMMHK-UHFFFAOYSA-N
Synonym 3-amino-2-naphthol, 2-naphthalenol, 3-amino, 2-amino-3-naphthol, 3-amino-2-hydroxynaphthalene, 2-naphthol, 3-amino, 2-amino-3-hydroxynaphthalene, unii-29bke27d17, 3-amino-naphthalen-2-ol, acmc-209ldx, 3-amino-2-naphthalenol
PubChem CID 79449
IUPAC Name 3-aminonaphthalen-2-ol
SMILES C1=CC=C2C=C(C(=CC2=C1)N)O
Specifications
Melting Point 232°C to 238°C
Odor Odorless
Quantity 25 g
Formula Weight 159.19
Percent Purity 97%
Chemical Name or Material 3-Amino-2-naphthol