missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-methyl-benzyl-d2 Alcohol, TRC

Product Code. 30326884
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30326884 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30326884 Supplier Toronto Research Chemicals Supplier No. TRCA614357100MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Amino-2-methyl-benzyl-d2 Alcohol
CAS 57414-76-3
Molecular Formula C8 2H2 H9 N O
Quantity 100 mg
Synonym Benzenemethan-d2-ol, 3-amino-2-methyl- (9CI), 3-Amino-2-methyl-benzyl-d2 alcohol, (3-Amino-2-methyl-phenyl)-dideuterio-methanol
InChI Formula InChI=1S/C8H11NO/c1-6-7(5-10)3-2-4-8(6)9/h2-4,10H,5,9H2,1H3/i5D2
SMILES [2H]C([2H])(O)c1cccc(N)c1C
IUPAC Name (3-amino-2-methylphenyl)-dideuteriomethanol
Molecular Weight (g/mol) 139.19
Formula Weight 139.0966
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.