Learn More
3-Amino-1,5-pentanediol, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Amino-1,5-pentanediol |
| CAS | 1117-23-3 |
| Molecular Formula | C5H13NO2 |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C5H13NO2/c6-5(1-3-7)2-4-8/h5,7-8H,1-4,6H2 |
| SMILES | NC(CCO)CCO |
| IUPAC Name | 3-aminopentane-1,5-diol |
| Molecular Weight (g/mol) | 119.16 |
| Formula Weight | 119.0946 |