Learn More
3-Acetyl-3-chlorodihydrofuranone, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Acetyl-3-chlorodihydrofuranone |
| CAS | 2986-00-7 |
| Molecular Formula | C6 H7 Cl O3 |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C6H7ClO3/c1-4(8)6(7)2-3-10-5(6)9/h2-3H2,1H3 |
| SMILES | CC(=O)C1(Cl)CCOC1=O |
| IUPAC Name | 3-acetyl-3-chlorooxolan-2-one |
| Molecular Weight (g/mol) | 162.57 |
| Formula Weight | 162.0084 |