missing translation for 'onlineSavingsMsg'
Learn More

3-Acetyl-2,5-dimethylthiophene, 99%

Product Code. 11403577
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11403577

Brand: Thermo Scientific Alfa Aesar A17329.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2530-10-1 | C9H22O3Si | 206.36 g/mol

It is used in the synthesis of heterocyclic ketimines, 3-acetyl-2,5-dimethylthiophene thiosemicarbazone and 3-acetyl-2,5- dimethylthiophene semicarbazone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of heterocyclic ketimines, 3-acetyl-2,5-dimethylthiophene thiosemicarbazone and 3-acetyl-2,5- dimethylthiophene semicarbazone.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2530-10-1
MDL Number MFCD00009763

Specifications

Density 1.086
Boiling Point 224°C
Flash Point 98°C (208°F)
Molecular Formula C8H10OS
Refractive Index 1.5455
Quantity 50 g
Beilstein 112095
Synonym 3-acetyl-2,5-dimethylthiophene, 1-2,5-dimethylthiophen-3-yl ethanone, ethanone, 1-2,5-dimethyl-3-thienyl, 2,5-dimethyl-3-thienyl methyl ketone, dimethylthienylcetone, 2,5-dimethyl-3-acetylthiophene, 1-2,5-dimethyl-3-thienyl ethanone, ketone, 2,5-dimethyl-3-thienyl methyl, 3-acetyl-2,5-dimethyl thiophene, unii-vdv865t91p
Solubility Information Soluble in alcohol. Insoluble in water.
InChI Key PUSJAEJRDNPYKM-UHFFFAOYSA-N
SMILES CC1=CC(=C(S1)C)C(=O)C
IUPAC Name 1-(2,5-dimethylthiophen-3-yl)ethanone
Molecular Weight (g/mol) 154.23
PubChem CID 520191
Formula Weight 154.23
Percent Purity 99%
Chemical Name or Material 3-Acetyl-2,5-dimethylthiophene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.