Learn More
3,8-Dinitro-6-phenyl-phenanthridine, TRC
Specifications
Specifications
| Chemical Name or Material | 3,8-Dinitro-6-phenyl-phenanthridine |
| CAS | 82921-86-6 |
| Molecular Formula | C19 H11 N3 O4 |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C19H11N3O4/c23-21(24)13-6-8-15-16-9-7-14(22(25)26)11-18(16)20-19(17(15)10-13)12-4-2-1-3-5-12/h1-11H |
| SMILES | [O-][N+](=O)c1ccc2c(c1)nc(c3ccccc3)c4cc(ccc24)[N+](=O)[O-] |
| IUPAC Name | 3,8-dinitro-6-phenylphenanthridine |
| Molecular Weight (g/mol) | 345.31 |
| Formula Weight | 345.075 |