Learn More
3,6-Octanedione, TRC
Product Code. 30135921
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135921 10 g 10g
30005935 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30135921 Supplier Toronto Research Chemicals Supplier No. TRCO23750010G
 Restricted Item
Estimated Shipment: 11-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2955-65-9
Molecular Formula C8 H14 O2
Molecular Weight (g/mol) 142.2
Synonym 3,6-Octanedione, 3,6-Octandione
IUPAC Name octane-3,6-dione
SMILES CCC(=O)CCC(=O)CC
Specifications
Quantity 10 g
InChI Formula InChI=1S/C8H14O2/c1-3-7(9)5-6-8(10)4-2/h3-6H2,1-2H3
Formula Weight 142.0994
Chemical Name or Material 3,6-Octanedione