Learn More
3,6-Dihydroxyflavone, 97%, Thermo Scientific Chemicals
Catalog Number 11481620
Click to view available options
Quantity:
1 g
5 g
108238-41-1
C15H10O4
254.241
MFCD00143082
XHLOLFKZCUCROE-UHFFFAOYSA-N
3,6-dihydroxyflavone, 3,6-dihydroxy-2-phenyl-4h-chromen-4-one, 6-hydroxyflavonol, 3,6-dihydroxy-flavone, 4h-1-benzopyran-4-one, 3,6-dihydroxy-2-phenyl, acmc-1c7fz, 3,6-dihydroxy-2-phenylchromone
688659
3,6-dihydroxy-2-phenylchromen-4-one
C1=CC=C(C=C1)C2=C(C(=O)C3=C(O2)C=CC(=C3)O)O
108238-41-1
C15H10O4
254.241
MFCD00143082
XHLOLFKZCUCROE-UHFFFAOYSA-N
3,6-dihydroxyflavone, 3,6-dihydroxy-2-phenyl-4h-chromen-4-one, 6-hydroxyflavonol, 3,6-dihydroxy-flavone, 4h-1-benzopyran-4-one, 3,6-dihydroxy-2-phenyl, acmc-1c7fz, 3,6-dihydroxy-2-phenylchromone
688659
3,6-dihydroxy-2-phenylchromen-4-one
C1=CC=C(C=C1)C2=C(C(=O)C3=C(O2)C=CC(=C3)O)O
3,6-Dihydroxyflavone,is used as an oxidative stress inducer - apoptosis inducer, it is widely used as a pharmaceutical intermediate and in cell survival applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,6-Dihydroxyflavone,is used as an oxidative stress inducer - apoptosis inducer, it is widely used as a pharmaceutical intermediate and in cell survival applications.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 231°C to 234°C
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 254.24
Percent Purity 97%
Chemical Name or Material 3,6-Dihydroxyflavone