Learn More
3,6-Dichloro-2-fluorobenzylamine, 97%, Thermo Scientific™
Product Code. 15404827
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15404827
Brand: Thermo Scientific Alfa Aesar H32132.MD
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 916420-66-1
Molecular Formula C7H6Cl2FN
Molecular Weight (g/mol) 194.03
MDL Number MFCD09025386
InChI Key CSAKSJHNYMDUQJ-UHFFFAOYSA-N
Synonym 3,6-dichloro-2-fluorobenzylamine, 1-3,6-dichloro-2-fluorophenyl methanamine, 3,6-dichloro-2-fluorophenyl methanamine, 3,6-dichloro-2-fluorophenyl methylamine
PubChem CID 46737552
IUPAC Name (3,6-dichloro-2-fluorophenyl)methanamine
SMILES C1=CC(=C(C(=C1Cl)CN)F)Cl
Specifications
Melting Point 44°C to 47°C
Quantity 250 mg
UN Number UN3259
Sensitivity Air Sensitive
Formula Weight 194.04
Percent Purity 97%
Chemical Name or Material 3,6-Dichloro-2-fluorobenzylamine