Learn More
3,6-Dibromo-9H-carbazole, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 3,6-Dibromo-9H-carbazole |
| CAS | 6825-20-3 |
| Molecular Formula | C12H7Br2N |
| Quantity | 1 g |
| Synonym | 3,6-Dibromocarbazole, NSC 121206 |
| InChI Formula | InChI=1S/C12H7Br2N/c13-7-1-3-11-9(5-7)10-6-8(14)2-4-12(10)15-11/h1-6,15H |
| SMILES | BrC1=CC=C2C(C(C=C(Br)C=C3)=C3N2)=C1 |
| IUPAC Name | 3,6-dibromo-9H-carbazole |
| Molecular Weight (g/mol) | 325 |
| Formula Weight | 322.89 |