missing translation for 'onlineSavingsMsg'
Learn More

3',5'-Dimethoxyacetophenone, 97%, ACROS Organics™

Product Code. 10307690 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10307690

Brand: Thermo Scientific Acros 171630010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Specifications

CAS 39151-19-4
CAS Min % 96.0
CAS Max % 100.0
Melting Point 36°C to 41°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C10H12O3
Linear Formula (CH3O)2C6H3COCH3
MDL Number MFCD00008739
Quantity 1 g
Beilstein 08, 274
Synonym 3',5'-dimethoxyacetophenone, 1-3,5-dimethoxyphenyl ethanone, 3,5-dimethoxyacetophenone, 1-3,5-dimethoxyphenyl ethan-1-one, ethanone, 1-3,5-dimethoxyphenyl, 1-acetyl-3,5-dimethoxybenzene, acetophenone, 3',5'-dimethoxy-6ci,7ci, acmc-20anci, pubchem3391, opera_id_1889
InChI Key YJKHOUIVWKQRSL-UHFFFAOYSA-N
SMILES COC1=CC(=CC(OC)=C1)C(C)=O
IUPAC Name 1-(3,5-dimethoxyphenyl)ethan-1-one
Molecular Weight (g/mol) 180.20
PubChem CID 95997
Formula Weight 180.2
Percent Purity ≥96.0%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 3', 5'-Dimethoxyacetophenone, 97%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.