Learn More
3,5-Difluoro-L-phenylalanine, TRC
Product Code. 30373396
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30373396 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30373396 Supplier Toronto Research Chemicals Supplier No. TRCD451205100MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3,5-Difluoro-L-phenylalanine
CAS 31105-91-6
Molecular Formula C9 H9 F2 N O2
Quantity 100 mg
InChI Formula InChI=1S/C9H9F2NO2/c10-6-1-5(2-7(11)4-6)3-8(12)9(13)14/h1-2,4,8H,3,12H2,(H,13,14)/t8-/m0/s1
SMILES N[C@@H](Cc1cc(F)cc(F)c1)C(=O)O
IUPAC Name (2S)-2-amino-3-(3,5-difluorophenyl)propanoic acid
Molecular Weight (g/mol) 201.17
Formula Weight 201.06