Learn More
3,5-Difluoro-2-methoxyaniline, 97%, Thermo Scientific™
Product Code. 15444457
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 15444457
Brand: Thermo Scientific Alfa Aesar H31816.03
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 41860-67-7
Molecular Formula C7H7F2NO
Molecular Weight (g/mol) 159.14
MDL Number MFCD09832386
InChI Key LJLOVRRJQKWWIU-UHFFFAOYSA-N
Synonym benzenamine, 3,5-difluoro-2-methoxy, 3,5-difluoro-2-methoxyphenylamine, benzenamine,3,5-difluoro-2-methoxy-9ci, benzenamine, 3,5-difluoro-2-methoxy-9ci
PubChem CID 20112223
IUPAC Name 3,5-difluoro-2-methoxyaniline
SMILES COC1=C(F)C=C(F)C=C1N
Specifications
Refractive Index 1.507
Quantity 1 g
UN Number UN2810
Formula Weight 159.14
Percent Purity 97%
Chemical Name or Material 3,5-Difluoro-2-methoxyaniline