Learn More
3,5-Dichloroaniline-2,4,6-d3, CDN
Product Code. 30321360
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30321360
Brand: CDN CDND70150.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3,5-Dichloroaniline D3 (Ring D3)
CAS 1219795-03-5
Molecular Formula C6 D3 H2 Cl2 N
Quantity 0.5 g
Synonym 3,5-Dichloroaniline D3 (Ring D3), Benzen-2,4,6-d3-amine, 3,5-dichloro-
InChI Formula InChI=1 S/C6H5Cl2N/c7-4-1-5(8)3-6(9)2-4/h1-3 H,9H2/i1D,2 D,3 D
SMILES [2 H]c1c(N)c([2 H])c(Cl)c([2 H])c1Cl
IUPAC Name 3,5-dichloro-2,4,6-trideuterioaniline
Molecular Weight (g/mol) 165.035
Formula Weight 163.999 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity