missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibenzyloxyacetophenone, 98%, ACROS Organics™

Product Code. 10522801 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10522801 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10522801 Supplier Thermo Scientific Acros Supplier No. 201330100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Specifications

CAS 28924-21-2
CAS Min % 97.5
CAS Max % 100.0
Melting Point 60°C to 64°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C22H20O3
Linear Formula (C6H5CH2O)2C6H3COCH3
MDL Number MFCD00004777
Quantity 10 g
Synonym 3',5'-dibenzyloxyacetophenone, 3,5-dibenzyloxyacetophenone, ethanone, 1-3,5-bis phenylmethoxy phenyl, 3,5-bis benzyloxy acetophenone, 1-3,5-bis phenylmethoxy phenyl ethanone, 1-3,5-bis benzyloxy phenyl ethanone, 3',5'-bis benzyloxy acetophenone, ethanone,1-3,5-bis phenylmethoxy phenyl
InChI Key KOJXGMJOTRYLBD-UHFFFAOYSA-N
SMILES CC(=O)C1=CC(=CC(=C1)OCC2=CC=CC=C2)OCC3=CC=CC=C3
IUPAC Name 1-[3,5-bis(phenylmethoxy)phenyl]ethanone
Molecular Weight (g/mol) 332.399
PubChem CID 120098
Formula Weight 332.39
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 3, 5-Dibenzyloxyacetophenone, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.