Learn More
3,5,6-Trichloro-4-hydroxy-2-picolinic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 3,5,6-Trichloro-4-hydroxy-2-picolinic Acid |
| CAS | 26449-73-0 |
| Molecular Formula | C6 H2 Cl3 N O3 |
| Quantity | 25 mg |
| Synonym | 3,5,6-Trichloro-4-hydroxy-2-pyridinecarboxylic Acid |
| InChI Formula | InChI=1S/C6H2Cl3NO3/c7-1-3(6(12)13)10-5(9)2(8)4(1)11/h(H,10,11)(H,12,13) |
| SMILES | OC(=O)c1nc(Cl)c(Cl)c(O)c1Cl |
| IUPAC Name | 3,5,6-trichloro-4-hydroxypyridine-2-carboxylic acid |
| Molecular Weight (g/mol) | 242.44 |
| Formula Weight | 240.91 |